Product Details

SNP ID
rs12405270
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:40376928 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAACTCTTTGGCAATGACTTTTA[G/T]TTTTAAATTCTTATACTTATTTAGG
Phenotype
MIM: 616916
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SMAP2 PubMed Links

Gene Details

Gene
SMAP2
Gene Name
small ArfGAP2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198978.1 Intron NP_001185907.1
NM_001198979.1 Intron NP_001185908.1
NM_001198980.1 Intron NP_001185909.1
NM_022733.2 Intron NP_073570.1
XM_011541960.1 Intron XP_011540262.1
XM_017002093.1 Intron XP_016857582.1

View Full Product Details