Product Details

SNP ID
rs11584399
Assay Type
Validated
NCBI dbSNP Submissions
40
Location
Chr.1:39523340 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GACCCACACCCAAACACGGACACAC[A/G]TGAACAGTCGCGTATGCATGCCTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BMP8A PubMed Links

Gene Details

Gene
BMP8A
Gene Name
bone morphogenetic protein 8a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181809.3 Intron NP_861525.2
XM_006710616.3 Intron XP_006710679.1
XM_011541381.2 Intron XP_011539683.1
XM_011541382.2 Intron XP_011539684.1
XM_017001198.1 Intron XP_016856687.1
Gene
OXCT2P1
Gene Name
3-oxoacid CoA-transferase 2 pseudogene 1
There are no transcripts associated with this gene.

Gene
PPIEL
Gene Name
peptidylprolyl isomerase E like pseudogene
There are no transcripts associated with this gene.

View Full Product Details