Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000070.2 | 7609 | Intron | NP_000061.1 | ||
NM_024344.1 | 7609 | Intron | NP_077320.1 | ||
NM_173087.1 | 7609 | Intron | NP_775110.1 | ||
NM_173088.1 | 7609 | Intron | NP_775111.1 | ||
NM_173089.1 | 7609 | Intron | NP_775112.1 | ||
NM_173090.1 | 7609 | Intron | NP_775113.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001284306.1 | 7609 | UTR 3 | NP_001271235.1 | ||
NM_001284307.1 | 7609 | UTR 3 | NP_001271236.1 | ||
NM_022473.2 | 7609 | UTR 3 | NP_071918.1 | ||
XM_005254591.2 | 7609 | UTR 3 | XP_005254648.1 | ||
XM_005254592.2 | 7609 | Intron | XP_005254649.1 | ||
XM_017022483.1 | 7609 | Intron | XP_016877972.1 | ||
XM_017022484.1 | 7609 | Intron | XP_016877973.1 | ||
XM_017022485.1 | 7609 | Intron | XP_016877974.1 |