Product Details

SNP ID
rs2296579
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:102481618 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TAGTACCCTGCCTCATCTGGGTATT[A/G]GAACTGCCTGCCTTTGGTGTGAAGA
Phenotype
MIM: 605143
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ACTR1A PubMed Links
Additional Information
For this assay, SNP(s) [rs117869402] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ACTR1A
Gene Name
ARP1 actin-related protein 1 homolog A, centractin alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005736.3 Intron NP_005727.1
Gene
MFSD13A
Gene Name
major facilitator superfamily domain containing 13A
There are no transcripts associated with this gene.

View Full Product Details