Product Details

SNP ID
rs12076250
Assay Type
Validated
NCBI dbSNP Submissions
53
Location
Chr.1:167053683 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGAGATGGGCCTGGAGGGACAGGTA[C/G]GATATTGACAGGCAGAGATGGAAAG
Phenotype
MIM: 602171
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GPA33 PubMed Links

Gene Details

Gene
GPA33
Gene Name
glycoprotein A33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005814.2 1783 UTR 3 NP_005805.1
XM_017000005.1 1783 UTR 3 XP_016855494.1

View Full Product Details