Product Details

SNP ID
hCV11342229
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:153544656 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACTCAGGCACTACCCACTCACCC[A/C]CAAGAAGCTGGGCAGCTCCCGGGTC
Phenotype
MIM: 176992 MIM: 114210 MIM: 176991 MIM: 114110
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC101928034 PubMed Links

Gene Details

Gene
LOC101928034
Gene Name
uncharacterized LOC101928034
There are no transcripts associated with this gene.

Gene
S100A3
Gene Name
S100 calcium binding protein A3
There are no transcripts associated with this gene.

Gene
S100A4
Gene Name
S100 calcium binding protein A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002961.2 257 Missense Mutation GGG,TGG G47W NP_002952.1
NM_019554.2 257 Missense Mutation GGG,TGG G47W NP_062427.1
Gene
S100A5
Gene Name
S100 calcium binding protein A5
There are no transcripts associated with this gene.

Gene
S100A6
Gene Name
S100 calcium binding protein A6
There are no transcripts associated with this gene.

View Full Product Details