Product Details

SNP ID
rs7281035
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:32785189 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCCATTGTAAAGGTCCTACTTGTA[A/G]TTAATAAGTAATCTGTGGAATGATG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C21orf62 PubMed Links

Gene Details

Gene
C21orf62
Gene Name
chromosome 21 open reading frame 62
There are no transcripts associated with this gene.

Gene
C21orf62-AS1
Gene Name
C21orf62 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details