Product Details

SNP ID
hCV11361351
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:10761367 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGTCTTTTCACCTTCGTAGGAG[C/G]TGTCACTCCAGGATGTCCGGAAAGA
Phenotype
MIM: 600280
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NUBP1 PubMed Links

Gene Details

Gene
NUBP1
Gene Name
nucleotide binding protein 1
There are no transcripts associated with this gene.

Gene
TVP23A
Gene Name
trans-golgi network vesicle protein 23 homolog A
There are no transcripts associated with this gene.

View Full Product Details