Product Details

SNP ID
rs14295
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:5044261 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCCTCCAGATGCCTTCTTCCCAG[G/T]ATGTGATCCGTGCCCTCCAGGATCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C16orf89 PubMed Links

Gene Details

Gene
C16orf89
Gene Name
chromosome 16 open reading frame 89
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098514.2 1331 UTR 3 NP_001091984.2
NM_152459.4 1331 UTR 3 NP_689672.4
XM_005255143.3 1331 Intron XP_005255200.2
XM_011522392.2 1331 UTR 3 XP_011520694.1
XM_017022972.1 1331 UTR 3 XP_016878461.1
XM_017022973.1 1331 Intron XP_016878462.1
XM_017022974.1 1331 Intron XP_016878463.1
Gene
NAGPA-AS1
Gene Name
NAGPA antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details