Product Details

SNP ID
rs1990265
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2786650 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGGAGAGTCCTGGCCCAGGGGCA[A/C]CAATCCTCACACCAGCCCGTCACCC
Phenotype
MIM: 613797 MIM: 600787
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PRSS33 PubMed Links

Gene Details

Gene
PRSS33
Gene Name
protease, serine 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152891.2 58 UTR 5 NP_690851.2
XM_011522451.2 58 Intron XP_011520753.1
XM_011522452.2 58 Intron XP_011520754.1
Gene
PRSS41
Gene Name
protease, serine 41
There are no transcripts associated with this gene.

Gene
TCEB2
Gene Name
transcription elongation factor B subunit 2
There are no transcripts associated with this gene.

View Full Product Details