Product Details

SNP ID
rs2030
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65996761 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATGGGAGATTCTACAGCACGCT[C/T]GACACTAACACATCACATCCATAAC
Phenotype
MIM: 603811
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BANF1 PubMed Links

Gene Details

Gene
BANF1
Gene Name
barrier to autointegration factor 1
There are no transcripts associated with this gene.

Gene
EIF1AD
Gene Name
eukaryotic translation initiation factor 1A domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242481.1 2480 UTR 3 NP_001229410.1
NM_001242482.1 2480 UTR 3 NP_001229411.1
NM_001242483.1 2480 UTR 3 NP_001229412.1
NM_001242484.1 2480 UTR 3 NP_001229413.1
NM_001242485.1 2480 UTR 3 NP_001229414.1
NM_001242486.1 2480 UTR 3 NP_001229415.1
NM_032325.3 2480 UTR 3 NP_115701.2
XM_017018412.1 2480 UTR 3 XP_016873901.1

View Full Product Details