Product Details

SNP ID
rs11339
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:106543371 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTCAGCAAAATCATTTATTTACA[C/G]AATGGGGAATGCTGGTTTGATTTTG
Phenotype
MIM: 614046 MIM: 600527
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARGLU1 PubMed Links

Gene Details

Gene
ARGLU1
Gene Name
arginine and glutamate rich 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018011.3 1694 UTR 3 NP_060481.3
Gene
EFNB2
Gene Name
ephrin B2
There are no transcripts associated with this gene.

View Full Product Details