Product Details

SNP ID
rs8527
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:182383585 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAATCCCAGATCACCATCTAAGAA[C/T]CACTTCCCTTGCTCTCTACTCAAAA
Phenotype
MIM: 138290
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLUL PubMed Links

Gene Details

Gene
GLUL
Gene Name
glutamate-ammonia ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033044.3 2546 UTR 3 NP_001028216.1
NM_001033056.3 2546 UTR 3 NP_001028228.1
NM_002065.6 2546 UTR 3 NP_002056.2
XM_006711278.1 2546 UTR 3 XP_006711341.1

View Full Product Details