Product Details

SNP ID
rs584088
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:78074714 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTATTTCCGAAAATGTATAAAACA[A/C]TAAAAATGAAAAATATTCCTATGTT
Phenotype
MIM: 603845 MIM: 601926
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NDUFC2 PubMed Links

Gene Details

Gene
NDUFC2
Gene Name
NADH:ubiquinone oxidoreductase subunit C2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204054.1 Intron NP_001190983.1
NM_001204055.1 Intron NP_001190984.1
NM_004549.5 Intron NP_004540.1
Gene
NDUFC2-KCTD14
Gene Name
NDUFC2-KCTD14 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001203260.1 Intron NP_001190189.1
NM_001203261.1 Intron NP_001190190.1
NM_001203262.1 Intron NP_001190191.1
Gene
THRSP
Gene Name
thyroid hormone responsive
There are no transcripts associated with this gene.

View Full Product Details