Product Details

SNP ID
rs2465520
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:203439491 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTCTCTTTTGGGGGGAGCAGGAGG[A/G]GGTCCTCTCCGCAACGTTGCATAGC
Phenotype
MIM: 606442 MIM: 609035
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABI2 PubMed Links

Gene Details

Gene
ABI2
Gene Name
abl-interactor 2
There are no transcripts associated with this gene.

Gene
RAPH1
Gene Name
Ras association (RalGDS/AF-6) and pleckstrin homology domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203365.3 3780 Intron NP_976241.1
NM_213589.2 3780 Silent Mutation CCC,CCT P1233P NP_998754.1
XM_005246777.3 3780 Silent Mutation CCC,CCT P1285P XP_005246834.1
XM_005246779.4 3780 Silent Mutation CCC,CCT P1285P XP_005246836.1
XM_006712695.3 3780 Silent Mutation CCC,CCT P1285P XP_006712758.1
XM_011511646.2 3780 Silent Mutation CCC,CCT P1285P XP_011509948.1
XM_011511647.1 3780 Silent Mutation CCC,CCT P1260P XP_011509949.1
XM_011511648.1 3780 Silent Mutation CCC,CCT P1258P XP_011509950.1
XM_011511649.2 3780 Intron XP_011509951.1

View Full Product Details