Product Details

SNP ID
rs35062851
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:26627308 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAGCCAGGGTAAACAGGGAGGAG[A/G]GATAGAAGAATCCTGTTTTTCAGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC3B PubMed Links

Gene Details

Gene
LRRC3B
Gene Name
leucine rich repeat containing 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317808.1 Intron NP_001304737.1
NM_001317809.1 Intron NP_001304738.1
NM_001317810.1 Intron NP_001304739.1
NM_001317811.1 Intron NP_001304740.1
NM_052953.3 Intron NP_443185.1
XM_005264849.3 Intron XP_005264906.1
XM_005264850.4 Intron XP_005264907.1
XM_011533332.2 Intron XP_011531634.1
XM_017005675.1 Intron XP_016861164.1

View Full Product Details