Product Details

SNP ID
rs2919879
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:88071133 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCTGTATGGTTTTGTGAAACTAAT[A/G]AAATGAATAAACTTTGTGTGTGTCT
Phenotype
MIM: 606846
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMYD1 PubMed Links

Gene Details

Gene
SMYD1
Gene Name
SET and MYND domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198274.3 Intron NP_938015.1
XM_005264156.2 Intron XP_005264213.1

View Full Product Details