Product Details

SNP ID
rs1986391
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:100297282 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGGAAGGGAATCCAACAGTCAGT[A/G]TATTTAAGAAGCCTTGGCCATATTT
Phenotype
MIM: 300460
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCDH19 PubMed Links

Gene Details

Gene
PCDH19
Gene Name
protocadherin 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105243.1 Intron NP_001098713.1
NM_001184880.1 Intron NP_001171809.1
NM_020766.2 Intron NP_065817.2
XM_011530997.2 Intron XP_011529299.1

View Full Product Details