Product Details

SNP ID
rs746035909
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:124365814 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTCCCAGACAGAAACAGGTTCTT[A/G]GAGTGCTGAGGTATAGTTGTATTTA
Phenotype
MIM: 300490 MIM: 300588
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SH2D1A PubMed Links

Gene Details

Gene
SH2D1A
Gene Name
SH2 domain containing 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114937.2 552 Nonsense Mutation TAG,TGG *64W NP_001108409.1
NM_002351.4 552 Nonsense Mutation TAG,TGG *64W NP_002342.1
Gene
TENM1
Gene Name
teneurin transmembrane protein 1
There are no transcripts associated with this gene.

View Full Product Details