Product Details

SNP ID
rs1032070
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:42466233 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTCCATTCCAGTAATGTTGCAGCAA[C/T]GAGGGAGGAAGTTTTTATCCTTAGC
Phenotype
MIM: 192130
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP6V0A1 PubMed Links

Gene Details

Gene
ATP6V0A1
Gene Name
ATPase H+ transporting V0 subunit a1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130020.1 Intron NP_001123492.1
NM_001130021.1 Intron NP_001123493.1
NM_005177.3 Intron NP_005168.2
XM_005257459.1 Intron XP_005257516.1
XM_005257461.1 Intron XP_005257518.1
XM_005257463.2 Intron XP_005257520.1
XM_011524908.1 Intron XP_011523210.1
XM_017024766.1 Intron XP_016880255.1
XM_017024767.1 Intron XP_016880256.1
XM_017024768.1 Intron XP_016880257.1
XM_017024769.1 Intron XP_016880258.1
XM_017024770.1 Intron XP_016880259.1
XM_017024771.1 Intron XP_016880260.1
XM_017024772.1 Intron XP_016880261.1
XM_017024773.1 Intron XP_016880262.1
XM_017024774.1 Intron XP_016880263.1

View Full Product Details