Product Details

SNP ID
rs1891107
Assay Type
Functionally tested
NCBI dbSNP Submissions
44
Location
Chr.1:226362741 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
TTAGGTTAAAGCCGAAACCCGTGCA[C/T]CTGGTTACTGGTGCCTGCACCAAGA
Phenotype
MIM: 173870
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PARP1 PubMed Links

Gene Details

Gene
PARP1
Gene Name
poly(ADP-ribose) polymerase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001618.3 Intron NP_001609.2

View Full Product Details