Product Details

SNP ID
rs9293642
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:75069201 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAGGAGTGATGATAAAGAGGGTT[C/T]CTTTTGGCTCTGCCAGTCTATGGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD31 PubMed Links

Gene Details

Gene
ANKRD31
Gene Name
ankyrin repeat domain 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164443.1 Intron NP_001157915.1
XM_011543298.2 Intron XP_011541600.1
XM_011543299.1 Intron XP_011541601.1
XM_011543300.2 Intron XP_011541602.1
XM_011543301.2 Intron XP_011541603.1
XM_011543302.2 Intron XP_011541604.1
XM_017009317.1 Intron XP_016864806.1
XM_017009318.1 Intron XP_016864807.1
XM_017009319.1 Intron XP_016864808.1
XM_017009320.1 Intron XP_016864809.1
XM_017009321.1 Intron XP_016864810.1
XM_017009322.1 Intron XP_016864811.1

View Full Product Details