Product Details
- SNP ID
-
rs2765148
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:23986659 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATGTTTAGAGGTGTGGATTTCTCA[A/C]CTTTAATTTATCTTTTTGGTTTGCT
- Phenotype
-
MIM: 613324
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SPATA13
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77971723] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SPATA13
- Gene Name
- spermatogenesis associated 13
View Full Product Details