Product Details

SNP ID
rs1127935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30657058 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACATAAATGATGCCCTCAATGGTG[A/G]GTGATGTCTCAGCAATGTTCGTTGC
Phenotype
MIM: 603405
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C6orf136 PubMed Links

Gene Details

Gene
C6orf136
Gene Name
chromosome 6 open reading frame 136
There are no transcripts associated with this gene.

Gene
DHX16
Gene Name
DEAH-box helicase 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164239.1 2462 Missense Mutation CCC,CTC P621L NP_001157711.1
NM_003587.4 2462 Missense Mutation CCC,CTC P681L NP_003578.2
XM_011514938.2 2462 Missense Mutation CCC,CTC P382L XP_011513240.1
XM_011514939.2 2462 Missense Mutation CCC,CTC P382L XP_011513241.1
XM_011514940.2 2462 Missense Mutation CCC,CTC P200L XP_011513242.1
XM_011514941.2 2462 Missense Mutation CCC,CTC P164L XP_011513243.1

View Full Product Details