Product Details

SNP ID
rs7635877
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:179596499 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CTGATTGAGAAAGTGCAGTACATAT[A/G]ACCTACTGGGCTGAAGTTCACCTCT
Phenotype
MIM: 604958 MIM: 611852 MIM: 603841
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACTL6A PubMed Links

Gene Details

Gene
ACTL6A
Gene Name
actin like 6A
There are no transcripts associated with this gene.

Gene
MRPL47
Gene Name
mitochondrial ribosomal protein L47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020409.2 Intron NP_065142.2
NM_177988.1 Intron NP_817125.1
Gene
NDUFB5
Gene Name
NADH:ubiquinone oxidoreductase subunit B5
There are no transcripts associated with this gene.

View Full Product Details