Product Details

SNP ID
rs12666341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:36391084 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATTACAGATTTCAGCAATTAGAG[G/T]TTACTTATATTCAAGCATGACATAA
Phenotype
MIM: 616027
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ANLN PubMed Links

Gene Details

Gene
ANLN
Gene Name
anillin actin binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284301.2 Intron NP_001271230.1
NM_001284302.2 Intron NP_001271231.1
NM_018685.4 Intron NP_061155.2
XM_006715746.1 Intron XP_006715809.1
XM_006715747.3 Intron XP_006715810.1
XM_017012354.1 Intron XP_016867843.1
XM_017012355.1 Intron XP_016867844.1
XM_017012356.1 Intron XP_016867845.1
Gene
KIAA0895
Gene Name
KIAA0895
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100425.1 Intron NP_001093895.1
NM_001199706.1 Intron NP_001186635.1
NM_001199707.1 Intron NP_001186636.1
NM_001199708.1 Intron NP_001186637.1
NM_001300956.1 Intron NP_001287885.1
NM_015314.2 Intron NP_056129.2
XM_005249689.3 Intron XP_005249746.1
XM_005249690.4 Intron XP_005249747.1

View Full Product Details