Product Details

SNP ID
rs4606052
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:63064118 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GGCTAATGATATAGATGGGCAGGTA[C/T]GAGGAAAGATGATAAAGCAATCCTT
Phenotype
MIM: 600415
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TTPA PubMed Links

Gene Details

Gene
TTPA
Gene Name
alpha tocopherol transfer protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000370.3 Intron NP_000361.1
XM_006716468.3 Intron XP_006716531.1

View Full Product Details