Product Details
- SNP ID
-
rs522587
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:10527720 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTAAGAGATACGTGTGTATGCATG[C/T]GTGTGTGTGAACGGACTTCTTGTTT
- Phenotype
-
MIM: 600429
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GCNT2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs141470231] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GCNT2
- Gene Name
- glucosaminyl (N-acetyl) transferase 2, I-branching enzyme (I blood group)
View Full Product Details