Product Details

SNP ID
rs14712
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:23891143 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTATATGCCAATCTACTTTGTTT[A/T]AAAAAGGTCTGAATCAGGACTTGTG
Phenotype
MIM: 604496 MIM: 602916
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NKIRAS1 PubMed Links

Gene Details

Gene
NKIRAS1
Gene Name
NFKB inhibitor interacting Ras like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020345.3 1297 Intron NP_065078.1
XM_005265076.3 1297 UTR 3 XP_005265133.1
XM_005265077.4 1297 UTR 3 XP_005265134.1
XM_005265078.3 1297 UTR 3 XP_005265135.1
Gene
UBE2E1
Gene Name
ubiquitin conjugating enzyme E2 E1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202476.1 1297 UTR 3 NP_001189405.1
NM_003341.4 1297 UTR 3 NP_003332.1
NM_182666.2 1297 UTR 3 NP_872607.1
XM_005265431.4 1297 Intron XP_005265488.1

View Full Product Details