Product Details

SNP ID
rs753659
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:133590934 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATTGCCCTTTGTGAAGGAAGCTTC[C/T]GGGGATCCTGAGAGAGGTACCAGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM163B PubMed Links

Gene Details

Gene
FAM163B
Gene Name
family with sequence similarity 163 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080515.2 Intron NP_001073984.1
XM_005272203.4 Intron XP_005272260.1
XM_005272204.4 Intron XP_005272261.1
XM_006717233.3 Intron XP_006717296.1
XM_011518912.2 Intron XP_011517214.1
XM_011518913.2 Intron XP_011517215.1
XM_017015017.1 Intron XP_016870506.1
XM_017015018.1 Intron XP_016870507.1

View Full Product Details