Product Details

SNP ID
rs13223
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:1633465 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTGGCTTATGAATGTTCTAATCA[A/G]GTGTGGAAGGAGCTGACACACGGAG
Phenotype
MIM: 605158
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PXDN PubMed Links

Gene Details

Gene
PXDN
Gene Name
peroxidasin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012293.2 5271 UTR 3 NP_036425.1
XM_005264707.3 5271 UTR 3 XP_005264764.1
XM_011510396.1 5271 UTR 3 XP_011508698.1
XM_011510397.2 5271 UTR 3 XP_011508699.1

View Full Product Details