Product Details

SNP ID
rs1884014
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:54398245 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAGAGCTAGAAAGAGCATTAGGTG[C/G]TTACAATTGCTAATCCCTATGTTGC
Phenotype
MIM: 123832
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
CDKN3 PubMed Links

Gene Details

Gene
CDKN3
Gene Name
cyclin dependent kinase inhibitor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130851.1 Intron NP_001124323.1
NM_005192.3 Intron NP_005183.2
XM_011536353.2 Intron XP_011534655.1

View Full Product Details