Product Details

SNP ID
rs1052879
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:179588962 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TCTTTCTCTCTAAATTTTCCTTCCG[G/T]GCTTTGATGCGGGCTCGTTTCTCAC
Phenotype
MIM: 604958 MIM: 611852
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ACTL6A PubMed Links

Gene Details

Gene
ACTL6A
Gene Name
actin like 6A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004301.4 539 Intron NP_004292.1
NM_177989.3 539 Intron NP_817126.1
NM_178042.3 539 Intron NP_829888.1
Gene
MRPL47
Gene Name
mitochondrial ribosomal protein L47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020409.2 539 Silent Mutation GCA,GCC A221A NP_065142.2
NM_177988.1 539 Silent Mutation GCA,GCC A111A NP_817125.1

View Full Product Details