Product Details

SNP ID
rs12730
Assay Type
Functionally Tested
NCBI dbSNP Submissions
39
Location
Chr.1:51908222 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATACAGAACATAGGAAGAAGAAAAA[A/C]GCATGAGAACATCTGCTTAGTTAGA
Phenotype
MIM: 179510
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
RAB3B PubMed Links
Additional Information
For this assay, SNP(s) [rs75682658] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
RAB3B
Gene Name
RAB3B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002867.3 12578 UTR 3 NP_002858.2
XM_017001958.1 12578 Intron XP_016857447.1

View Full Product Details