Product Details

SNP ID
rs1806158
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:31817240 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTTTACGCGCTGCCCAGCTCTGT[C/G]GGCCGCATTCTTTCGTACTCCGCGT
Phenotype
MIM: 607108
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DKFZp686K1684 PubMed Links

Gene Details

Gene
DKFZp686K1684
Gene Name
uncharacterized LOC440034
There are no transcripts associated with this gene.

Gene
PAUPAR
Gene Name
PAX6 upstream antisense RNA
There are no transcripts associated with this gene.

Gene
PAX6
Gene Name
paired box 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000280.4 Intron NP_000271.1
NM_001127612.1 Intron NP_001121084.1
NM_001258462.1 Intron NP_001245391.1
NM_001258463.1 Intron NP_001245392.1
NM_001258464.1 Intron NP_001245393.1
NM_001258465.1 Intron NP_001245394.1
NM_001310158.1 Intron NP_001297087.1
NM_001310159.1 Intron NP_001297088.1
NM_001310160.1 Intron NP_001297089.1
NM_001310161.1 Intron NP_001297090.1
NM_001604.5 Intron NP_001595.2

View Full Product Details