Product Details

SNP ID
rs2816606
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:105249697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGACACGGTGCTGGCCACTCTGTA[C/T]GCTGCTAAGAAGTACATCGTCCCAG
Phenotype
MIM: 604902
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BRF1 PubMed Links

Gene Details

Gene
BRF1
Gene Name
BRF1, RNA polymerase III transcription initiation factor 90 kDa subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242786.1 Intron NP_001229715.1
NM_001242787.1 Intron NP_001229716.1
NM_001242788.1 Intron NP_001229717.1
NM_001242789.1 Intron NP_001229718.1
NM_001242790.1 Intron NP_001229719.1
NM_001519.3 Intron NP_001510.2
NM_145685.2 Intron NP_663718.1
XM_005267561.3 Intron XP_005267618.1
XM_005267563.3 Intron XP_005267620.1
XM_006720123.2 Intron XP_006720186.1
XM_011536672.2 Intron XP_011534974.1
XM_011536673.1 Intron XP_011534975.1
XM_011536674.1 Intron XP_011534976.1
Gene
BTBD6
Gene Name
BTB domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033271.2 Intron NP_150374.2
XM_005268188.3 Intron XP_005268245.1

View Full Product Details