Product Details

SNP ID
rs2070314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:33032729 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTACTATTCCCCTTTCAAGTACCA[A/G]CTGATCTGTCACCTCCTCCTGGAAG
Phenotype
MIM: 614108 MIM: 614110
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BPIFB2 PubMed Links

Gene Details

Gene
BPIFB2
Gene Name
BPI fold containing family B member 2
There are no transcripts associated with this gene.

Gene
BPIFB6
Gene Name
BPI fold containing family B member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174897.2 Intron NP_777557.1
XM_017027663.1 Intron XP_016883152.1

View Full Product Details