Product Details

SNP ID
rs2074613
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140334979 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCACCTCATTTACGTGATCCTCA[C/T]AACATCCTTATGAGGTAGATGGACA
Phenotype
MIM: 126150
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HBEGF PubMed Links

Gene Details

Gene
HBEGF
Gene Name
heparin binding EGF like growth factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001945.2 Intron NP_001936.1

View Full Product Details