Product Details

SNP ID
rs2279380
Assay Type
Functionally tested
NCBI dbSNP Submissions
61
Location
Chr.1:114091606 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TGAAAATGCAGCCGCAGTCCCACCA[C/T]GCCTCCTCCTTTTCCAGGGTTGTGG
Phenotype
MIM: 607718
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SYT6 PubMed Links

Gene Details

Gene
SYT6
Gene Name
synaptotagmin 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270805.1 2056 UTR 3 NP_001257734.1
NM_205848.3 2056 UTR 3 NP_995320.1
XM_017000371.1 2056 UTR 3 XP_016855860.1

View Full Product Details