Product Details
- SNP ID
-
rs2267433
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:41481768 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTTGAAAGCACAATCCAGAAGGCC[A/G]GTGGTCGTGTCACGGCTCTGCCTTC
- Phenotype
-
MIM: 100850
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ACO2
PubMed Links
Gene Details
- Gene
- ACO2
- Gene Name
- aconitase 2
View Full Product Details