Product Details
- SNP ID
-
rs2254675
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:120970431 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGATGGTCTGTTTCCATGATGTGA[A/G]TACCTCCAGCTGGGCCGAACTGACA
- Phenotype
-
MIM: 142410
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HNF1A
PubMed Links
Gene Details
- Gene
- HNF1A
- Gene Name
- HNF1 homeobox A
There are no transcripts associated with this gene.
- Gene
- HNF1A-AS1
- Gene Name
- HNF1A antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details