Product Details

SNP ID
rs2290423
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:9428830 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGTAGCAGTGTCTGTCTGTGTTTT[G/T]CATTCAGATCTTGCTATCCACACAA
Phenotype
MIM: 605586
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IPO7 PubMed Links

Gene Details

Gene
IPO7
Gene Name
importin 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006391.2 Intron NP_006382.1
Gene
SNORA23
Gene Name
small nucleolar RNA, H/ACA box 23
There are no transcripts associated with this gene.

View Full Product Details