Product Details

SNP ID
rs2210641
Assay Type
Validated
NCBI dbSNP Submissions
45
Location
Chr.1:86425496 on Build GRCh38
Set Membership
JSNP Validated
Context Sequence [VIC/FAM]
GAAAAGGTAAGAATCCAGGATTTCA[C/T]TCAATGATCTCAAGGGGAAAAAAAA
Phenotype
MIM: 604003
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CLCA2 PubMed Links

Gene Details

Gene
CLCA2
Gene Name
chloride channel accessory 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006536.5 Intron NP_006527.1
XM_011542448.2 Intron XP_011540750.1

View Full Product Details