Product Details

SNP ID
hCV15942062
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:131601385 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAAGTATATCAGATTAGCTTTAA[A/C]AATAACTTTATTACAGTTCCTATCT
Phenotype
MIM: 611266
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
POTEI PubMed Links

Gene Details

Gene
POTEI
Gene Name
POTE ankyrin domain family member I
There are no transcripts associated with this gene.

Gene
POTEKP
Gene Name
POTE ankyrin domain family member K, pseudogene
There are no transcripts associated with this gene.

View Full Product Details