Product Details

SNP ID
rs2233465
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:184244046 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCTGCCAAGGCTCATTCCCAATG[A/G]CCAGCCTTCATGCTACTCATTGAGG
Phenotype
MIM: 608750 MIM: 610145 MIM: 611620
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALG3 PubMed Links

Gene Details

Gene
ALG3
Gene Name
ALG3, alpha-1,3- mannosyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006941.2 Intron NP_001006942.1
NM_005787.5 Intron NP_005778.1
XM_011512322.1 Intron XP_011510624.1
XM_011512323.2 Intron XP_011510625.1
Gene
ECE2
Gene Name
endothelin converting enzyme 2
There are no transcripts associated with this gene.

Gene
MIR1224
Gene Name
microRNA 1224
There are no transcripts associated with this gene.

Gene
VWA5B2
Gene Name
von Willebrand factor A domain containing 5B2
There are no transcripts associated with this gene.

View Full Product Details