Product Details
- SNP ID
-
rs2272470
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:54233306 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGAAGAAAAAACAGTTATGTCTTTT[C/T]TCCCCCTTTGGATTTACTTTGGATT
- Phenotype
-
MIM: 604478
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CBX5
PubMed Links
Gene Details
- Gene
- CBX5
- Gene Name
- chromobox 5
- Gene
- MIR3198-2
- Gene Name
- microRNA 3198-2
There are no transcripts associated with this gene.
View Full Product Details