Product Details

SNP ID
rs2305340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:7113370 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAATGAAGAAGTTTTTGTCCTTGC[A/G]TATAAACTTAGGCCAGGCATGGGTC
Phenotype
MIM: 608974 MIM: 611866
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1RL PubMed Links

Gene Details

Gene
C1RL
Gene Name
complement C1r subcomponent like
There are no transcripts associated with this gene.

Gene
C1RL-AS1
Gene Name
C1RL antisense RNA 1
There are no transcripts associated with this gene.

Gene
RBP5
Gene Name
retinol binding protein 5
There are no transcripts associated with this gene.

View Full Product Details