Product Details

SNP ID
rs3020727
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:86789062 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGACATGTGCGCGCGCGCTGCG[A/G]GCTTCGTCGACGGAACACACCCGCT
Phenotype
MIM: 186910
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD8A PubMed Links

Gene Details

Gene
CD8A
Gene Name
CD8a molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145873.1 Intron NP_001139345.1
NM_001768.6 Intron NP_001759.3
NM_171827.3 Intron NP_741969.1

View Full Product Details