Product Details

SNP ID
rs2363242
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:21294548 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCCTCAGGCTGTTGCTGTTTGGAC[A/G]TTGGAGCCAGACAGACCTGCCTTTA
Phenotype
MIM: 602315
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAP2K3 PubMed Links

Gene Details

Gene
MAP2K3
Gene Name
mitogen-activated protein kinase kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316332.1 Intron NP_001303261.1
NM_002756.4 Intron NP_002747.2
NM_145109.2 Intron NP_659731.1
XM_005256723.2 Intron XP_005256780.1
XM_011523958.2 Intron XP_011522260.1
XM_011523959.1 Intron XP_011522261.1
XM_017024857.1 Intron XP_016880346.1
XM_017024858.1 Intron XP_016880347.1
XM_017024859.1 Intron XP_016880348.1

View Full Product Details