Product Details
- SNP ID
-
rs2110964
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:37252816 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGATGCTAGCTTGACTTGTTTTTC[A/C]TTAAAAAAACAAACAAACATATATT
- Phenotype
-
MIM: 615898
MIM: 607077
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NDUFAF7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139159885] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NDUFAF7
- Gene Name
- NADH:ubiquinone oxidoreductase complex assembly factor 7
- Gene
- PRKD3
- Gene Name
- protein kinase D3
View Full Product Details